Skip to Content
Merck
All Photos(1)

Key Documents

241342

Sigma-Aldrich

1,2-Epoxybutane

≥99%

Synonym(s):

α-Butylene oxide, 1,2-Butylene oxide, 1-Butene oxide, Ethyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102411
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

2.2 (vs air)

vapor pressure

140 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

698 °F

expl. lim.

19 %

refractive index

n20/D 1.384 (lit.)

bp

63 °C (lit.)

density

0.829 g/mL at 25 °C (lit.)

SMILES string

CCC1CO1

InChI

1S/C4H8O/c1-2-4-3-5-4/h4H,2-3H2,1H3

InChI key

RBACIKXCRWGCBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 109975-ALDRICH | 1,2-Epoxybutane 99%

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yuan Sun et al.
Carbohydrate research, 344(15), 1999-2004 (2009-08-26)
A new soluble cyclodextrin derivative 6-O-(2-hydroxybutyl)-beta-cyclodextrin (6-HB-beta-CD) was prepared. Its molecular binding and recognition ability were investigated with the comparison of beta-cyclodextrin (beta-CD), 2-O-(2-hydroxypropyl)-beta-cyclodextrin (2-HP-beta-CD), 6-O-(2-hydroxypropyl)-beta-cyclodextrin (6-HP-beta-CD), and 2-O-(2-hydroxybutyl)-beta-cyclodextrin (2-HB-beta-CD). The relationship between the complex stability constants and the possible
M Katz et al.
Journal of environmental pathology and toxicology, 3(5-6), 171-187 (1980-06-01)
Nitrosopiperidine, sodium nitrite and 1,2 epoxybutane were tested in the Ames agar incorporation assay in an attempt to establish exact criteria for detecting the activity of these weak mutagens. As regards minimum concentrations it was determined that at 500 microgram
T Nakamura et al.
Biochemical and biophysical research communications, 180(1), 124-130 (1991-10-15)
Halohydrin hydrogen-halide-lyase, which catalyzes the interconversion of halohydrins to epoxides, purified from a recombinant E. coli was found to catalyze the transformation of 1,2-epoxybutane into beta-hydroxyvaleronitrile in the presence of cyanide. Chloride inhibited competitively the formation of beta-hydroxyvaleronitrile. The enzyme
A M Rossi et al.
Teratogenesis, carcinogenesis, and mutagenesis, 3(1), 75-87 (1983-01-01)
In vivo and in vitro methodologies that have employed the yeast Schizosaccharomyces pombe as genetic indicator have been utilized to investigate the mutagenicity of two trichloroethylene (TCE) samples of pure and technical grade. Mutagenicity assays were also performed on two
D Weinstein et al.
Environmental mutagenesis, 3(1), 1-9 (1981-01-01)
Based on the findings of Nagao et al [1978] that phenacetin is negative in the standard Ames test with Aroclor induced rat S-9 and positive with hamster S-9, the test was performed with a mixture of rat/hamster S-9. Phenacetin was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service