Skip to Content
Merck
All Photos(1)

Documents

238848

Sigma-Aldrich

Salicylaldehyde hydrazone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CH=NNH2
CAS Number:
Molecular Weight:
136.15
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

98%

mp

97-98 °C (lit.)

SMILES string

N\N=C\c1ccccc1O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Wang et al.
Free radical biology & medicine, 164, 369-380 (2021-01-16)
Metastasis is the major cause of death in women with advanced ovarian cancer. Epithelial ovarian cancer cells can dissociate directly from extracellular matrix (ECM) and form spheroids to spread through the peritoneal cavity. Loss of ECM hinders the survival of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service