Skip to Content
Merck
All Photos(1)

Documents

234486

Sigma-Aldrich

3-(Dimethylamino)propionitrile

98%

Synonym(s):

DMAPN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2CN
CAS Number:
Molecular Weight:
98.15
Beilstein:
773779
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.426 (lit.)

bp

171 °C/750 mmHg (lit.)

mp

−43 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCC#N

InChI

1S/C5H10N2/c1-7(2)5-3-4-6/h3,5H2,1-2H3

InChI key

MTPJEFOSTIKRSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogenation of 3-(dimethylamino)propionitrile over palladium catalysts has been investigated.

Application

3-(Dimethylamino)propionitrile has been used:
  • to initiate the organic polymerization during the synthesis of class I hybrid poly(N-isopropylacrylamide)/silica hydrogels
  • in preparation of cysteine modifying agents based on the solvatochromic fluorophore 4-N,N-dimethylamino-1,8-naphthalimide (4-DMN)
Catalyst similar to TEMED used to form oxygen free radicals from ammonium persulfate. Free radicals cause acrylamide and bis-acrylamide to polymerize.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galen Loving et al.
Bioconjugate chemistry, 20(11), 2133-2141 (2009-10-14)
The solvatochromic fluorophore 4-N,N-dimethylamino-1,8-naphthalimide (4-DMN) possesses extremely sensitive emission properties due largely to the low intrinsic fluorescence it exhibits in polar protic solvents such as water. This makes it well suited as a probe for the detection of a wide
Hydrogenation of 3-(dimethylamino) propionitrile over palladium catalysts.
Krupka J, et al.
Collection of Czechoslovak Chemical Communications, 65(11), 1805-1819 (2000)
P Banet et al.
The journal of physical chemistry. B, 113(45), 14914-14919 (2009-11-06)
Class I hybrid poly(N-isopropylacrylamide)/silica hydrogels, PNIPAM/SiO2, were prepared by a new one shot synthesis. In this approach, the free-radical polymerization of vinyl groups of N-isopropylacrylamide (NIPAM) and the hydrolysis-condensation of alkoxy groups of tetramethoxysilane (TMOS) are performed concomitantly using sodium
R J Jaeger et al.
Journal of environmental pathology and toxicology, 4(2-3), 555-562 (1980-09-01)
The toxicity of the mixture NIAX-ESN which contains 95% dimethylaminopropionitrile was evaluated in male rats. Large doses (2.0 ml/kg ip) rapidly produced CNS excitation followed by depression and death. The mixture (given orally as two doses of 0.31 or 0.62
Occupationally related neurologic abnormalities--Massachusetts.
MMWR. Morbidity and mortality weekly report, 30(30), 265-266 (1981-08-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service