Skip to Content
Merck
All Photos(1)

Key Documents

229792

Sigma-Aldrich

Potassium bromide

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050405
PubChem Substance ID:

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

Assay

99.99% trace metals basis

form

solid

impurities

≤150.0 ppm Trace Metal Analysis

refractive index

n20/D 1.5598 (lit.)

bp

1435 °C/1 atm (lit.)

mp

734 °C (lit.)

solubility

ethanol: slightly soluble(lit.)

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hao-Long Zhou et al.
Nature communications, 6, 6917-6917 (2015-04-22)
The dynamic behaviours of host frameworks and guest molecules have received much attention for their great relevance with smart materials, but little has been developed to control or understand the host-guest interplay. Here we show that the confined guest can
Zai-Chao Ma et al.
Marine biotechnology (New York, N.Y.), 17(4), 511-522 (2015-05-20)
In this study, in order to directly and efficiently convert inulin into pullulan, the INU1 gene from Kluyveromyces maximum KM was integrated into the genomic DNA and actively expressed in the high pullulan producer Aureobasidium melanogenum P16 isolated from the
Sophie Rodrigues et al.
Probiotics and antimicrobial proteins, 7(1), 45-51 (2014-10-22)
Vibrio tapetis CECT4600 is a pathogenic Gram-negative bacterium causing the brown ring disease in the Manila clam Ruditapes philippinarum. This vibriosis is induced by bacterial attachment on the periostracal lamina, yielding a decalcification of the bivalve shell. As in many
Jun Pan et al.
Organic letters, 13(18), 4974-4976 (2011-08-26)
A facile Pd-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) is described. This method allows convenient access to a variety of aryl, heteroaryl, and vinyl halides in good to excellent yields and with
Igor L Dalinger et al.
Chemistry, an Asian journal, 10(9), 1987-1996 (2015-06-23)
A new family of energetic compounds, nitropyrazoles bearing a trinitromethyl moiety at the nitrogen atom of the heterocycle, was designed. The desirable high-energy dense oxidizers 3,4-dinitro- and 3,5-dinitro-1-(trinitromethyl)pyrazoles were synthesized in good yields by destructive nitration of the corresponding 1-acetonylpyrazoles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service