Skip to Content
Merck
All Photos(2)

Key Documents

229504

Sigma-Aldrich

Cadmium carbonate

powder and chunks, ≥99.996% trace metals basis

Synonym(s):

Otavite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdCO3
CAS Number:
Molecular Weight:
172.42
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.996% trace metals basis
form:
powder and chunks
solubility:
H2O: insoluble(lit.)
aqueous acid: slightly soluble(lit.)

Assay

≥99.996% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤40.0 ppm Trace Metal Analysis

solubility

H2O: insoluble(lit.)
aqueous acid: slightly soluble(lit.)

density

4.258 g/mL at 25 °C (lit.)

SMILES string

[Cd++].[O-]C([O-])=O

InChI

1S/CH2O3.Cd/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

GKDXQAKPHKQZSC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1

Target Organs

Lungs,Kidney,Bone

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kian Siong et al.
Journal of hazardous materials, 167(1-3), 1237-1241 (2009-02-24)
We studied the relationship between macro-algae Chara (Stoneworts) calcite (CaCO(3)) encrustation and the speciation of cadmium (Cd) accumulated by the plant. Results showed that 17% of the total Cd (0.3mgkg(-1)) accumulated by Chara fibrosa exposed to 1 microg Cd L(-1)
E Suzuki et al.
Steroids, 60(3), 277-284 (1995-03-01)
The synthesis of 15-N-acetylglucosaminides of 15 alpha-hydroxyesterone, 15 alpha-hydroxyestradiol, and 15 alpha-hydroxyestriol (estetrol) is described. The latter two were prepared by condensation of 2-acetamido-1 alpha-chloro-1,2-dideoxy-3,4,6-trio-O-acetyl-D-glucopyranose with appropriately protected 15 alpha-hydroxyestrogens by the Koenigs-Knorr reaction employing cadmium carbonate as a catalyst.
V Vĕtvicka et al.
Folia biologica, 31(1), 20-33 (1985-01-01)
In the high-responder A/J mice a lower percentage of peritoneal macrophages express Fc receptors and accordingly phagocytize less intensely via these receptors than the macrophages of the low-IgG-responder C57BL/10ScSn strain. Similarly, the phagocytosis mediated by the receptors for foreign substances
Haidong Yu et al.
Journal of the American Chemical Society, 129(8), 2333-2337 (2007-02-06)
In this article, we have demonstrated one-step solid-phase transformation from high-quality cadmium carbonate microcrystals into highly nanoporous cadmium oxide. The high crystal quality of cadmium carbonate is critical for the successful fabrication of porous nanoarchitectures with predetermined morphology and well-controlled
[Micromethod for the determination of polymorphonuclear granulocyte phagocytic ability using cadmium carbonate crystals].
U Thielebeule et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 29(2), 69-70 (1983-02-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service