Skip to Content
Merck
All Photos(1)

Documents

229261

Sigma-Aldrich

6,7-Dimethoxy-2-tetralone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

87-89 °C (lit.)

SMILES string

COc1cc2CCC(=O)Cc2cc1OC

Gene Information

human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Application

For enamine preparation and elaboration to dopamine agonists.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W K Brewster et al.
Journal of medicinal chemistry, 33(6), 1756-1764 (1990-06-01)
trans-10,11-Dihydroxy-5,6,6a,7,8,12b-hexahydrobenzo[a]phenan thridine (4a, dihydrexidine) has been found to be a highly potent and selective agonist of the dopamine D1 receptor in rat brain. Dihydrexidine had an EC50 of approximately 70 nM in activating dopamine-sensitive rat striatal adenylate cyclase and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service