Skip to Content
Merck
All Photos(1)

Key Documents

229199

Sigma-Aldrich

Diphenylmercury(II)

Synonym(s):

Mercury(II)diphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2Hg
CAS Number:
Molecular Weight:
354.80
Beilstein:
637234
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: mercury
reagent type: catalyst

mp

121-124 °C (lit.)

density

2.32 g/mL at 25 °C (lit.)

SMILES string

[Hg](c1ccccc1)c2ccccc2

InChI

1S/2C6H5.Hg/c2*1-2-4-6-5-3-1;/h2*1-5H;

InChI key

HWMTUNCVVYPZHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service