Skip to Content
Merck
All Photos(1)

Key Documents

226432

Sigma-Aldrich

Methyltriphenoxyphosphonium iodide

96%

Synonym(s):

Triphenyl phosphite methoiodide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CHF 44.70
50 G
CHF 219.00

CHF 44.70


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
CHF 44.70
50 G
CHF 219.00

About This Item

Linear Formula:
CH3P(I)(OC6H5)3
CAS Number:
Molecular Weight:
452.22
Beilstein:
3580189
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

CHF 44.70


Estimated to ship on17 April 2025


Request a Bulk Order

Quality Level

Assay

96%

mp

142-146 °C (lit.)

SMILES string

[I-].C[P+](Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C19H18O3P.HI/c1-23(20-17-11-5-2-6-12-17,21-18-13-7-3-8-14-18)22-19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1

InChI key

VKTOBGBZBCELGC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used in the transformation of the 5′-hydroxyl group in nucleosides to the 5′-deoxy-5′-iodo group.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 1795-1795 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service