Skip to Content
Merck
All Photos(1)

Key Documents

226297

Sigma-Aldrich

(−)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol

97%

Synonym(s):

(−)-2,3-O-Isopropylidene-1,4-di-O-tosyl-L-threitol, (−)-2,3-O-Isopropylidene-L-threitol 1,4-ditosylate, (4S,5S)-4,5-Bis(tosyloxymethyl)-2,2-dimethyl-1,3-dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O8S2
CAS Number:
Molecular Weight:
470.56
Beilstein:
99610
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]23/D −12.3°, c = 8.8 in chloroform

mp

90-92 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OC[C@@H]2OC(C)(C)O[C@H]2COS(=O)(=O)c3ccc(C)cc3

InChI

1S/C21H26O8S2/c1-15-5-9-17(10-6-15)30(22,23)26-13-19-20(29-21(3,4)28-19)14-27-31(24,25)18-11-7-16(2)8-12-18/h5-12,19-20H,13-14H2,1-4H3/t19-,20-/m0/s1

InChI key

KPFDKWNWYAXRNJ-PMACEKPBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service