220620
1,3-Dioxane-5,5-dimethanol
90%, technical grade
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
grade
technical grade
Assay
90%
form
solid
mp
58-60 °C (lit.)
SMILES string
OCC1(CO)COCOC1
InChI
1S/C6H12O4/c7-1-6(2-8)3-9-5-10-4-6/h7-8H,1-5H2
InChI key
ZTISUHQLYYPYFA-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service