Skip to Content
Merck
All Photos(1)

Documents

220396

Sigma-Aldrich

Ethyl 2-acetoxy-2-methylacetoacetate

97%

Synonym(s):

Ethyl 2-acetyllactate acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC(O2CCH3)(CH3)CO2C2H5
CAS Number:
Molecular Weight:
202.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

80-84 °C/1 mmHg (lit.)

density

1.079 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)(OC(C)=O)C(C)=O

InChI

1S/C9H14O5/c1-5-13-8(12)9(4,6(2)10)14-7(3)11/h5H2,1-4H3

InChI key

RHFWCEYBHLGCKH-UHFFFAOYSA-N

Application

Ethyl 2-acetoxy-2-methylacetoacetate was used to prepare 2-acetolactate via hydrolysis with NaOH. It was also used in isolation and characterization of a novel thermophilic bacterium Geobacillus sp. for the production of acetoin and 2,3-butanediol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zijun Xiao et al.
Biotechnology for biofuels, 5(1), 88-88 (2012-12-12)
Acetoin and 2,3-butanediol are two important biorefinery platform chemicals. They are currently fermented below 40°C using mesophilic strains, but the processes often suffer from bacterial contamination. This work reports the isolation and identification of a novel aerobic Geobacillus strain XT15
Lingxue Lu et al.
Microbial cell factories, 19(1), 102-102 (2020-05-14)
Acetoin, especially the optically pure (3S)- or (3R)-enantiomer, is a high-value-added bio-based platform chemical and important potential pharmaceutical intermediate. Over the past decades, intense efforts have been devoted to the production of acetoin through green biotechniques. However, efficient and economical
Chin-Yu Chen et al.
Scientific reports, 8(1), 7176-7176 (2018-05-10)
Ketol-acid reductoisomerase (KARI) is a bifunctional enzyme in the second step of branched-chain amino acids biosynthetic pathway. Most KARIs prefer NADPH as a cofactor. However, KARI with a preference for NADH is desirable in industrial applications including anaerobic fermentation for
R Y Xing et al.
Journal of bacteriology, 173(6), 2086-2092 (1991-03-01)
Methanococcus aeolicus, Methanococcus maripaludis, and Methanococcus voltae contain similar levels of four enzymes of branched-chain amino acid biosynthesis: acetohydroxy acid synthase, acetohydroxy acid isomeroreductase, dihydroxy acid dehydratase, and transaminase B. Following growth at low partial pressures of H2-CO2, the levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service