Skip to Content
Merck
All Photos(1)

Key Documents

218820

Sigma-Aldrich

Phenoxathiin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8OS
CAS Number:
Molecular Weight:
200.26
Beilstein:
143232
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

bp

150-152 °C/5 mmHg (lit.)

mp

52-56 °C (lit.)

SMILES string

O1c2ccccc2Sc3ccccc13

InChI

1S/C12H8OS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8H

InChI key

GJSGGHOYGKMUPT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurica Varlan et al.
Molecules (Basel, Switzerland), 15(6), 3905-3919 (2010-07-27)
The interactions of 3-carboxyphenoxathiin with Bovine Serum Albumin (BSA) and Human Serum Albumin (HSA) have been studied by fluorescence and circular dichroism spectroscopy. The binding of 3-carboxyphenoxathiin quenches the BSA and HSA fluorescence, revealing a 1:1 interaction with a binding
A selective, reversible, competitive inhibitor of monoamine oxidase A containing no nitrogen, with negligible potentiation of tyramine-induced blood pressure rise.
M Harfenist et al.
Journal of medicinal chemistry, 34(9), 2931-2933 (1991-09-01)
L J Fitzgerald et al.
Acta crystallographica. Section C, Crystal structure communications, 47 ( Pt 2), 381-385 (1991-02-15)
Mr = 200.25, orthorhombic, P2(1)2(1)2(1), a = 7.758(2), b = 20.506(3), c = 5.896(2) A, V = 938.0(4) A3, Z = 4, Dx = 1.42 g cm-3, lambda(Mo K alpha) = 0.71069 A, mu = 2.88 cm-1, F(000) = 416
H Nojiri et al.
Journal of bacteriology, 181(10), 3105-3113 (1999-05-13)
Carbazole 1,9a-dioxygenase (CARDO) from Pseudomonas sp. strain CA10 is a multicomponent enzyme that catalyzes the angular dioxygenation of carbazole, dibenzofuran, and dibenzo-p-dioxin. It was revealed by gas chromatography-mass spectrometry and 1H and 13C nuclear magnetic resonance analyses that xanthene and
K Stolze et al.
Chemico-biological interactions, 77(3), 283-289 (1991-01-01)
Several derivatives of the phenothiazine cation radicals intercalated into DNA have been investigated using a new flow orientation technique. The anisotropic hyperfine coupling constants of both the parallel and the perpendicular orientation relative to the magnetic field were measured and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service