Skip to Content
Merck
All Photos(1)

Key Documents

217050

Sigma-Aldrich

2-Methoxy-1,3-dioxolane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
Beilstein:
103177
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

129-130 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COC1OCCO1

InChI

1S/C4H8O3/c1-5-4-6-2-3-7-4/h4H,2-3H2,1H3

InChI key

VRAYTNFBRROPJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxy-1,3-dioxolane was used in the synthesis of :
  • [4,5-bis(hydroxymethyl)-1,3-dioxolan-2-yl]nucleosides, potential inhibitors of HIV
  • diastereoisomeric cyclic acetals

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jonas Brånalt et al.
The Journal of organic chemistry, 61(11), 3599-3603 (1996-05-31)
The synthesis of 1,3-dioxolan-2-ylnucleosides and related chemistry is described. We have shown that 2-methoxy-1,3-dioxolane (6) reacts with silylated thymine and trimethylsilyl triflate to give the acyclic formate ester 1-[2-(formyloxy)ethyl]thymine (8) rather than 1-(1,3-dioxolan-2-yl)thymine (7). A tentative mechanism which could explain
Synthesis of highly substituted 2, 6-anti-configured tetrahydropyrans. First steps towards an efficient access to amphidinol 3 ring system.
Dubost C, et al.
Tetrahedron Letters, 46(23), 4005-4009 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service