Skip to Content
Merck
All Photos(1)

Key Documents

216119

Sigma-Aldrich

Diethyl 3,3′-(phenethylimino)dipropionate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2N(CH2CH2CO2C2H5)2
CAS Number:
Molecular Weight:
321.41
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical grade

Assay

90%

refractive index

n20/D 1.495 (lit.)

bp

190-193 °C/2 mmHg (lit.)

density

1.032 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCN(CCC(=O)OCC)CCc1ccccc1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service