Skip to Content
Merck
All Photos(2)

Documents

213764

Sigma-Aldrich

2-Phenyl-1,2-propanediol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H5)(OH)CH2OH
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

bp

160-162 °C/26 mmHg (lit.)

mp

44-45 °C (lit.)

SMILES string

CC(O)(CO)c1ccccc1

InChI

1S/C9H12O2/c1-9(11,7-10)8-5-3-2-4-6-8/h2-6,10-11H,7H2,1H3

InChI key

LNCZPZFNQQFXPT-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Goroncy-Bermes et al.
Zentralblatt fur Hygiene und Umweltmedizin = International journal of hygiene and environmental medicine, 195(5-6), 366-376 (1994-06-01)
Phenoxypropanols have been used for some time as ingredients in surface and instrument disinfectants. Since the residual solutions of these preparations are discharged into the waste-water, the biodegradability of the ingredients used is of great importance. Tests by means of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service