Skip to Content
Merck
All Photos(1)

Key Documents

210692

Sigma-Aldrich

Acid Blue 45

Dye content 50 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N2Na2O10S2
CAS Number:
Molecular Weight:
474.33
Colour Index Number:
63010
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Pricing and availability is not currently available.

composition

Dye content, 50%

mp

>300 °C (lit.)

λmax

595 nm

SMILES string

[Na+].[Na+].Nc1cc(c(O)c2C(=O)c3c(N)cc(c(O)c3C(=O)c12)S([O-])(=O)=O)S([O-])(=O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Ensafi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(9), 1739-1743 (2001-08-17)
A sensitive, rapid and selective procedure is proposed for the flow injection determinations of periodate by spectrofluorometric detection. The method is based on the reaction of periodate with Alizarin Navy Blue in basic solution. The reagents and manifold variables influence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service