Skip to Content
Merck
All Photos(1)

Key Documents

19980

Sigma-Aldrich

Butylhydrazine oxalate salt

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NHNH2 · (COOH)2
CAS Number:
Molecular Weight:
178.19
Beilstein:
3704357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

mp

160-165 °C

solubility

H2O: soluble 0.1 g/10 mL, clear, colorless

SMILES string

CCCCNN.OC(=O)C(O)=O

InChI

1S/C4H12N2.C2H2O4/c1-2-3-4-6-5;3-1(4)2(5)6/h6H,2-5H2,1H3;(H,3,4)(H,5,6)

InChI key

WXOBLSRQYOWIBF-UHFFFAOYSA-N

Application

Butylhydrazine oxalate salt may be used in the chemical synthesis studies.

Other Notes

Sales restrictions may apply

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service