Skip to Content
Merck
All Photos(1)

Key Documents

197033

Sigma-Aldrich

3-(Pyrrol-1-ylmethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2
CAS Number:
Molecular Weight:
158.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

59-64 °C (lit.)

SMILES string

C(c1cccnc1)n2cccc2

InChI

1S/C10H10N2/c1-2-7-12(6-1)9-10-4-3-5-11-8-10/h1-8H,9H2

InChI key

PJVWPGGKILHMKW-UHFFFAOYSA-N

Application

3-(Pyrrol-1-ylmethyl)pyridine (pmp) was used in the synthesis of:
  • electronically conducting copolymer films of [Ru(2,2′-bipyridine)2 (3-{pyrrol-1-ylmethyl} pyridine)Cl]+[1]
  • electronically conductive anion exchange polymers[2]
  • 1-methyl-3-(pyrrol-1-ylmethyl)pyridinium (MPP) iodide, used for preparation of poly[1-methyl-3-(pyrrol-1-ylmethyl)pyridinium] chloride films[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electronically conductive anion exchange polymers based on polypyrrole: Preparation, characterization, electrostatic binding of ferrocyanide and electrocatalysis of ascorbic acid oxidation.
Mao H and Pickup PG.
J. Electroanal. Chem. Interfac. Electrochem., 265(1), 127-142 (1989)
Current enhancing effect of poly [1-methyl-3-(pyrrol-1-ylmethyl) pyridinium] films on the electrode reaction of anthraquinonedisulfonate.
Komura T, et al.
Journal of Electroanalytical Chemistry, 478(1), 9-16 (1999)
Conducting polypyrrole films containing [Ru (2, 2'-bipyridine)2(3-{pyrrol-1-ylmethyl} pyridine) Cl]+: Electrochemistry, spectroelectrochemistry, electronic conductivity, and ionic conductivity.
Ochmanska J and Pickup PG.
J. Electroanal. Chem. Interfac. Electrochem., 271(1), 83-105 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service