Skip to Content
Merck
All Photos(1)

Key Documents

193798

Sigma-Aldrich

3-(Trifluoromethyl)phenylacetone

97%

Synonym(s):

1-(α,α,α-Trifluoro-m-tolyl)-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2COCH3
CAS Number:
Molecular Weight:
202.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.457 (lit.)

bp

89-90 °C/0.5 mmHg (lit.)

density

1.204 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Cc1cccc(c1)C(F)(F)F

InChI

1S/C10H9F3O/c1-7(14)5-8-3-2-4-9(6-8)10(11,12)13/h2-4,6H,5H2,1H3

InChI key

JPHQCDCEBDRIOL-UHFFFAOYSA-N

Application

3-(Trifluoromethyl)phenylacetone was used in the synthesis of:
  • (±)-[1-(3-trifluoromethyl)phenyl]-2-propylamine via reductive amination reaction[1]
  • N-{2-[1-methyl-2-(3-trifluoromethylphenyl]}-aminoethanol[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic one-pot resolution of two nucleophiles: alcohol and amine.
Garci'a-Urdiales E, et al.
Tetrahedron Asymmetry, 11(7), 1459-1463 (2000)
New prenylamine-analogues: investigations of their influence on calcium-dependent biological systems.
Caldirola P, et al.
European Journal of Medicinal Chemistry, 28(7), 555-568 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service