Skip to Content
Merck
All Photos(2)

Key Documents

192961

Sigma-Aldrich

4-Hydroxy-7-trifluoromethyl-3-quinolinecarboxylic acid

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6F3NO3
CAS Number:
Molecular Weight:
257.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

mp

259-260 °C (dec.) (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1cnc2cc(ccc2c1O)C(F)(F)F

InChI

1S/C11H6F3NO3/c12-11(13,14)5-1-2-6-8(3-5)15-4-7(9(6)16)10(17)18/h1-4H,(H,15,16)(H,17,18)

InChI key

BIRIVPOTERXIOW-UHFFFAOYSA-N

Gene Information

human ... CSNK2A1(1457)

Application

4-Hydroxy-7-trifluoromethyl-3-quinolinecarboxylic acid was used in the preparation of (4-hydroxy-7-trifluoromethylquinolin-3-yl)formaldehyde[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P B Koneru et al.
Pharmaceutical research, 10(4), 515-520 (1993-04-01)
A series of eight new N-hydroxy-N'-aminoguanidine (HAG) Schiff bases [ArCH = NNHC(= NH)NHOH.tosylate] was synthesized as potential antitumor agents through the inhibition of the enzyme ribonucleotide reductase (EC 1.17.4.1). Five of the HAG derivatives (LK02 through LK06) were designed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service