Skip to Content
Merck
All Photos(1)

Documents

186309

Sigma-Aldrich

Chlorosulfonic acid

99%

Synonym(s):

Sulfuric chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClSO3H
CAS Number:
Molecular Weight:
116.52
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4 (vs air)

vapor pressure

1 mmHg ( 25 °C)
3.3 mmHg ( 37.7 °C)

Assay

99%

form

liquid

expl. lim.

37.7 %

refractive index

n20/D 1.433 (lit.)

pKa 

-6

bp

151-152 °C/755 mmHg (lit.)

density

1.753 g/mL at 25 °C (lit.)

SMILES string

OS(Cl)(=O)=O

InChI

1S/ClHO3S/c1-5(2,3)4/h(H,2,3,4)

InChI key

XTHPWXDJESJLNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

SPE tube or plate

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart D Chambers et al.
Journal of chromatography. A, 1218(2), 263-269 (2010-12-15)
Carbon-clad zirconia particles have been converted into ion exchange media through addition of charged latexes after covalent modification of the carbon surface. A variety of methodologies were investigated to introduce a negative charge to the carbon surface in the form
Shimona Geresh et al.
Journal of biochemical and biophysical methods, 50(2-3), 179-187 (2001-12-14)
Polysaccharides are natural polymers with a variety of properties that may be translated into significant commercial applications. A program of chemical modifications of the extracellular polysaccharides of red microalgae, such as Porphyridium sp. and Rhodella reticulata, has been undertaken by
Natnael Behabtu et al.
Nature nanotechnology, 5(6), 406-411 (2010-06-01)
Graphene combines unique electronic properties and surprising quantum effects with outstanding thermal and mechanical properties. Many potential applications, including electronics and nanocomposites, require that graphene be dispersed and processed in a fluid phase. Here, we show that graphite spontaneously exfoliates
Yongxu Sun et al.
International journal of biological macromolecules, 44(1), 14-17 (2008-10-28)
In this study, three chemically sulfated polysaccharides (SPAPs) were derived from one water-soluble polysaccharide (PAP) of Polyporus albicans mycelia by chlorosulfonic acid-pyridine method. The effects of polysaccharides on the immune function were examined after the mice were intragastrical administrated with
Fang Han et al.
International journal of biological macromolecules, 36(4), 201-207 (2005-08-17)
A polysaccharide YCP was prepared from a marine filamentous fungus Keissleriella sp. YS4108, which exhibited as a molecular weight (Mw) of 2.4x10(3) kDa and its three sulfated derivatives (YCP-SL, YCP-SM and YCP-SH) were synthesized, the degree of substitution (DS) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service