Skip to Content
Merck
All Photos(2)

Key Documents

186082

Sigma-Aldrich

2-Methyl-2-propene-1-sulfonic acid sodium salt

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CH2SO3Na
CAS Number:
Molecular Weight:
158.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

>300 °C (lit.)

functional group

sulfonic acid

SMILES string

[Na+].CC(=C)CS([O-])(=O)=O

InChI

1S/C4H8O3S.Na/c1-4(2)3-8(5,6)7;/h1,3H2,2H3,(H,5,6,7);/q;+1/p-1

InChI key

SZHIIIPPJJXYRY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-2-propene-1-sulfonic acid sodium salt was used as anionic monomer to investigate the effect of different cationic or anionic moieties in acrylonitrile-based copolymers on the interaction with fibroblasts[1]. It was used to reduce the homopolymerization during graft copolymerization of methyl methacrylate with water-emulsified solution onto preirradiated jute fiber[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Cheung et al.
Blood purification, 9(3), 153-163 (1991-01-01)
Protein adsorption occurs upon blood exposure to hemodialysis membranes. While adsorption of certain proteins may be beneficial, adsorption of others may be undesirable. In the present study, we investigated the binding of recombinant human erythropoietin (rHuEPO) to different types of
Arantxazu Isla et al.
Journal of clinical pharmacology, 45(11), 1294-1304 (2005-10-22)
The pharmacokinetics of meropenem were characterized in 20 patients with different degrees of renal function who underwent continuous renal replacement therapy. Previously, no differences were detected in vitro in the removal of meropenem by continuous venovenous hemofiltration or continuous venovenous
A Gutierrez et al.
Blood purification, 12(2), 95-105 (1994-01-01)
The effect of blood-membrane interaction on several biocompatibility parameters has been investigated. Four groups of healthy subjects underwent sham hemodialysis, i.e. the establishment of blood-dialysis membrane contact, but without circulating dialysate, using cellulose-based membranes (regenerated cellulose and cellulose acetate) or
Graft copolymerization reaction of water-emulsified methyl methacrylate with preirradiated jute fiber.
Khan F and Ahmad SR.
Journal of Applied Polymer Science, 65(3), 459-468 (1997)
Minoru Furuta et al.
ASAIO journal (American Society for Artificial Internal Organs : 1992), 57(4), 293-299 (2011-04-19)
Many maintenance hemodialysis (MHD) patients have recently been treated with high flux (HF) dialysis membranes such as polysulfone (PSu) membranes. However, the appropriateness of HF for elderly MHD remains to be elucidated. In order to estimate hemodialysis (HD) efficiency, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service