Skip to Content
Merck
All Photos(1)

Documents

184837

Sigma-Aldrich

2-Amino-3-methyl-1-butanol

97%

Synonym(s):

DL-Valinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CH2OH
CAS Number:
Molecular Weight:
103.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.4543 (lit.)

bp

75-77 °C/8 mmHg (lit.)

density

0.936 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(N)CO

InChI

1S/C5H13NO/c1-4(2)5(6)3-7/h4-5,7H,3,6H2,1-2H3

InChI key

NWYYWIJOWOLJNR-UHFFFAOYSA-N

Application

2-Amino-3-methyl-1-butanol was used in the synthesis of 1-allyl-2-pyrroleimines. It was also used as chiral nucleophile in the total synthesis of benanomicin-pradimicin antibiotics. It was used as homochiral guest compound during synthesis of crown ethers containing two chiral subunits.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of homochiral phenolic crown ethers containing para-substituted phenol moiety and chiral subunits derived from (S)-1-phenylethane-1, 2-diol: their chiral recognition behaviour in complexation with neutral amines.
Naemura K, et al.
Tetrahedron Asymmetry, 8(6), 873-882 (1997)
Vincenzo Giulio Albano et al.
The Journal of organic chemistry, 73(21), 8376-8381 (2008-10-02)
1-Allyl-2-pyrroleimines obtained from (S)-valinol and (S)-phenylglycinol underwent highly diastereoselective addition of allylmagnesium chloride, used in excess amounts, to give the corresponding secondary amines with concomitant allyl to (Z)-1-propenyl isomerization of the 1-pyrrole substituent. Transformation of the 2-amino alcohol moiety to
Andrei V Malkov et al.
Chemical communications (Cambridge, England), (15)(15), 1948-1949 (2003-08-23)
New N,P-ligands 4-6, derived from valinol and prolinol, respectively, have been developed for the asymmetric, copper(I)-catalysed conjugate addition of diethylzinc to unsaturated ketones; the tertiary amide group has been shown to effectively relay the chiral information from the ligand backbone
E Spitzer et al.
Biochemistry international, 14(4), 581-588 (1987-04-01)
A procedure for purification of plasma membranes from bovine mammary gland has been developed. The binding capacity of EGF to the plasma membranes from mammary tissue of pregnant cows was equal to 335 fmol per mg of protein thus being
Minoru Tamiya et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(35), 9791-9823 (2007-10-02)
A general approach to the regio- and stereoselective total synthesis of the benanomicin-pradimicin antibiotics (BpAs) is described. Construction of the aglycon has been achieved by 1) the diastereoselective ring-opening of a biaryl lactone by using (R)-valinol as a chiral nucleophile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service