Skip to Content
Merck
All Photos(1)

Key Documents

183393

Sigma-Aldrich

Boron tris(trifluoroacetate) solution

1.0 M in trifluoroacetic acid

Synonym(s):

BTFA, Tris[trifluoroacetoxy]borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO2)3B
CAS Number:
Molecular Weight:
349.86
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

form

liquid

concentration

1.0 M in trifluoroacetic acid

density

1.521 g/mL at 25 °C

SMILES string

FC(F)(F)C(=O)OB(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F

InChI

1S/C6BF9O6/c8-4(9,10)1(17)20-7(21-2(18)5(11,12)13)22-3(19)6(14,15)16

InChI key

CWBHKBKGKCDGDM-UHFFFAOYSA-N

Application

Reactant for:
  • Trifluoroacetylation of arenes

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service