Skip to Content
Merck
All Photos(1)

Key Documents

18069

Sigma-Aldrich

(R)-(+)-1-(4-Bromophenyl)ethylamine

≥96.0% (sum of enantiomers, GC)

Synonym(s):

(R)-(+)-4-Bromo-α-methylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
Beilstein:
2412319
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (sum of enantiomers, GC)

optical activity

[α]20/D +20.5±1°, c = 3% in methanol

refractive index

n20/D 1.566

density

1.390 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](N)c1ccc(Br)cc1

InChI

1S/C8H10BrN/c1-6(10)7-2-4-8(9)5-3-7/h2-6H,10H2,1H3/t6-/m1/s1

InChI key

SOZMSEPDYJGBEK-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service