17347
2-Bromo-5-hydroxy-4-methoxybenzaldehyde
technical, ≥90% (HPLC)
Synonym(s):
5-Bromoisovanillin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H7BrO3
CAS Number:
Molecular Weight:
231.04
Beilstein:
1942858
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Assay
≥90% (HPLC)
form
solid
mp
≥95 °C (lit.)
≥95 °C
SMILES string
COc1cc(Br)c(C=O)cc1O
InChI
1S/C8H7BrO3/c1-12-8-3-6(9)5(4-10)2-7(8)11/h2-4,11H,1H3
InChI key
AHYSXUDLJOFNAB-UHFFFAOYSA-N
Application
2-Bromo-5-hydroxy-4-methoxybenzaldehyde was used in the synthesis of 4-bromo-2-methoxy-5-(2-methoxyethenyl)phenol.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of (?)-6H-benzofuro [3a, 3, 2, ef][3] benzazepine: an unnatural analog of (−)-galanthamine.
Poschalko A, et al.
Tetrahedron, 58(8), 1513-1518 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service