Skip to Content
Merck
All Photos(1)

Key Documents

17336

Sigma-Aldrich

4-(2-Fmoc-hydrazino)benzoic acid

≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N2O4
CAS Number:
Molecular Weight:
374.39
Beilstein:
8082614
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥95.0% (HPLC)

reaction suitability

reaction type: solution phase peptide synthesis
reagent type: linker

impurities

≤1% water
≤5% residual solvent

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(NNC(=O)OCC2c3ccccc3-c4ccccc24)cc1

InChI

1S/C22H18N2O4/c25-21(26)14-9-11-15(12-10-14)23-24-22(27)28-13-20-18-7-3-1-5-16(18)17-6-2-4-8-19(17)20/h1-12,20,23H,13H2,(H,24,27)(H,25,26)

InChI key

MLVJMSRGLQCZDE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Filippo Buono et al.
Cells, 9(7) (2020-07-24)
Genetic cardiomyopathies are characterized by changes in the function and structure of the myocardium. The development of a novel in vitro model could help to better emulate healthy and diseased human heart conditions and may improve the understanding of disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service