Skip to Content
Merck
All Photos(1)

Key Documents

172758

Sigma-Aldrich

2,3,5-Tribromothiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4HBr3S
CAS Number:
Molecular Weight:
320.83
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.664 (lit.)

bp

120 °C/11 mmHg (lit.)

mp

25-28 °C (lit.)

density

2.483 g/mL at 25 °C (lit.)

SMILES string

Brc1cc(Br)c(Br)s1

InChI

1S/C4HBr3S/c5-2-1-3(6)8-4(2)7/h1H

InChI key

SKDNDSLDRLEELJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,5-Tribromothiophene was used in the synthesis of 2,5-bis(dicyanomethylene)-3-bromo-2,5-dihydrothiophene[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the reaction of 2, 5-dihalothiophenes with tetracyanoethylene oxide.
Gronowitz SALO and UPPSTROM B.
Acta Chemica Scandinavica. Series B, 28, 981-985 (1974)
Khalil K Hussain et al.
Biosensors & bioelectronics, 155, 112094-112094 (2020-02-25)
A robust amperometric sensor was developed for the lactate detection in the extracellular matrix of cancer cells. The sensor was fabricated by separately immobilizing nicotinamide adenine dinucleotide (NAD+) onto a carboxylic acid group and lactate dehydrogenase (LDH) onto an amine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service