Skip to Content
Merck
All Photos(1)

Key Documents

162558

Sigma-Aldrich

4-Fluoro-2-nitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)NH2
CAS Number:
Molecular Weight:
156.11
Beilstein:
2210197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

90-94 °C (lit.)

functional group

fluoro
nitro

SMILES string

Nc1ccc(F)cc1[N+]([O-])=O

InChI

1S/C6H5FN2O2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H,8H2

InChI key

PUGDHSSOXPHLPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Fluoro-2-nitroaniline forms complexes with cobalt(II), nickel(II) and copper(II)[1].

Application

4-Fluoro-2-nitroaniline was used as starting reagent in the synthesis of 4-fluoro-N-ethyl-2-nitroaniline[2] and N-(4-fluoro-2-nitrophenyl)-β-alanine[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Hussenether et al.
Bioorganic & medicinal chemistry, 12(10), 2625-2637 (2004-04-28)
Novel 4-arylpiperazin-1-yl-substituted 2,3-dihydro-1H-1,4- and 1H-1,5-benzodiazepines and their aza-analogues were synthesized as debenzoclozapine derivatives for evaluation as potential D4-ligands. While Ki values of some of the title compounds came within the range of clozapine, they showed an impressively greater selectivity over
S Singh et al.
The Journal of pharmacology and experimental therapeutics, 296(2), 600-611 (2001-02-13)
The diseases of cystic fibrosis (CF) and chronic obstructive pulmonary disease (COPD) are characterized by mucus-congested airways. Agents that stimulate the secretion of Cl- are anticipated to facilitate mucociliary clearance and thus be of benefit in the treatment of CF
Complexes of copper (II), nickel (II) and cobalt (II) with 2-fluoro, 5-nitroaniline and 4-fluoro, 2-nitroaniline.
DeVoto G, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 38(7), 725-728 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service