Skip to Content
Merck
All Photos(2)

Documents

160075

Sigma-Aldrich

1-Phenyl-1-cyclopropanecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C3H4CO2H
CAS Number:
Molecular Weight:
162.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

85-87 °C (lit.)

SMILES string

OC(=O)C1(CC1)c2ccccc2

InChI

1S/C10H10O2/c11-9(12)10(6-7-10)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)

InChI key

IWWCCNVRNHTGLV-UHFFFAOYSA-N

Related Categories

Application

1-Phenyl-1-cyclopropanecarboxylic acid was used in the preparation of bis[(1-phenyl-1-cyclopropyl)carbonyl] peroxide via reaction with hydrogen peroxide in the presence of N,N′-dicyclohexylcarbodiimide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis [(1-phenyl-1-cyclopropyl) carbonyl] peroxide.
Bethell D, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 38(5), 1677-1679 (1982)
Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service