Skip to Content
Merck
All Photos(2)

Key Documents

157279

Sigma-Aldrich

Ethyl 2-formyl-1-cyclopropanecarboxylate, predominantly trans

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC3H4CO2C2H5
CAS Number:
Molecular Weight:
142.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

60-65 °C/0.6 mmHg (lit.)

density

1.074 g/mL at 25 °C (lit.)

functional group

aldehyde
ester

storage temp.

2-8°C

SMILES string

CCOC(=O)C1CC1C=O

InChI

1S/C7H10O3/c1-2-10-7(9)6-3-5(6)4-8/h4-6H,2-3H2,1H3

InChI key

MDWXTLNIZCHBJE-UHFFFAOYSA-N

General description

Ethyl 2-formyl-1-cyclopropanecarboxylate was formed by acid catalyzed reaction between acrolein and ethyl diazoacetate.[1]

Application

Ethyl 2-formyl-1-cyclopropanecarboxylate was used in the preparation of ethyl trans-2-(4-(methylphenyl)sulfonyl-4,5-dihydrooxazol-5-yl)cyclopropanecarboxylate.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid catalyzed reactions of a, ?-unsaturated aldehydes and ethyl diazoacetate.
Branstetter B and Hossain MM.
Tetrahedron Letters, 47(2), 221-223 (2006)
I J De Esch et al.
Journal of medicinal chemistry, 42(7), 1115-1122 (1999-04-10)
Various approaches to the synthesis of all four stereoisomers of 2-(1H-imidazol-4-yl)cyclopropylamine (cyclopropylhistamine) are described. The rapid and convenient synthesis and resolution of trans-cyclopropylhistamine is reported. The absolute configuration of its enantiomers was determined by single-crystal X-ray crystallographic analysis. The distinct

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service