Skip to Content
Merck
All Photos(1)

Key Documents

15689

Sigma-Aldrich

1-Hexyl-3-methylimidazolium chloride

dry, ≥98.5% (HPLC)

Synonym(s):

HMIMCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19ClN2
CAS Number:
Molecular Weight:
202.72
Beilstein:
8356525
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (HPLC)

form

liquid

concentration

50%±1% in acetonitrile

impurities

≤1000 ppm water

SMILES string

[Cl-].CCCCCCn1cc[n+](C)c1

InChI

1S/C10H19N2.ClH/c1-3-4-5-6-7-12-9-8-11(2)10-12;/h8-10H,3-7H2,1-2H3;1H/q+1;/p-1

InChI key

NKRASMXHSQKLHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service