Skip to Content
Merck
All Photos(1)

Key Documents

156787

Sigma-Aldrich

4-Chloro-DL-phenylalanine ethyl ester hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CH(NH2)CO2C2H5·HCl
CAS Number:
Molecular Weight:
264.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

168-170 °C (lit.)

SMILES string

Cl.CCOC(=O)C(N)Cc1ccc(Cl)cc1

InChI

1S/C11H14ClNO2.ClH/c1-2-15-11(14)10(13)7-8-3-5-9(12)6-4-8;/h3-6,10H,2,7,13H2,1H3;1H

InChI key

FDQIKGOQYNZYOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Ciarlone
Journal of dental research, 60(2), 182-186 (1981-02-01)
Lidocaine and procaine seizure thresholds were studied. The i.p. median convulsant dose (CD50) of lidocaine and procaine with saline pre-treatment was 95 and 240 mg/kg, respectively. Dopamine depletion by pre-treatment with d1-alpha-methyl-p-tyrosine plus dihydroxyphenylserine resulted in a significant drop of
Rebeca Santos Marques de Carvalho et al.
Behavioural brain research, 221(1), 75-82 (2011-03-08)
Baicalein (BA), one of the main flavonoids obtained from the Chinese medicinal herb Scutellaria baicalensis, usually exerts several pharmacological effects. In the central nervous system (CNS), BA exerts a protective effect on neurons against several neuronal insults among other effects
Joel Putnam et al.
Journal of chromatography. A, 1218(37), 6302-6307 (2011-07-30)
Acid/base mobile phase modifiers affect enantioseparations in ways that are not fully understood yet, for the lack of systematic studies. This makes chiral analysis of some pharmaceuticals difficult to reproduce. Once a column has been exposed to a modifier, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service