All Photos(1)
About This Item
Linear Formula:
ClC6H4COCH2CH2Cl
CAS Number:
Molecular Weight:
203.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
95%
bp
135-137 °C/0.6 mmHg (lit.)
mp
48-51 °C (lit.)
SMILES string
ClCCC(=O)c1ccc(Cl)cc1
InChI
1S/C9H8Cl2O/c10-6-5-9(12)7-1-3-8(11)4-2-7/h1-4H,5-6H2
InChI key
AYFJBHFMQODYBC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
177.8 °F - closed cup
Flash Point(C)
81 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jose Luis Gama Flores et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 54(13), 1311-1317 (2019-08-09)
Cladocerans are constantly exposed to humic substances in nature, yet the effects of these substances on their survival and reproduction are not well known. Here, the effects of humic substances (20 and 40 mg L-1) (HS) on the life history variables
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service