Skip to Content
Merck
All Photos(1)

Documents

140929

Sigma-Aldrich

D-(−)-Fructose

98%

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
12352201

Assay

98%

optical activity

[α]25/D −89.0°, c = 5 in H2O

mp

119-122 °C (dec.) (lit.)

solubility

alcohol: soluble 1 gm in 15 ml
methanol: soluble 1 gm in 14 ml
H2O: freely soluble
acetone: freely soluble (Hot)
acetone: slightly soluble (Cold)
pyridine: soluble

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(-)-Fructose has been used as a test compound utilized in NMR study for examining the mechanism of dehydration of D-fructose to 5-hydroxymethylfurfural in DMSO at 150°C.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ananda S Amarasekara et al.
Carbohydrate research, 343(18), 3021-3024 (2008-10-03)
The anomeric composition of d-fructose in dimethyl sulfoxide changes when the solution is heated from room temperature to 150 degrees C, with a small increase in the alpha-furanose form at the expense of the beta-pyranose tautomer. Additionally, a small amount
M Ledochowski et al.
Scandinavian journal of gastroenterology, 35(10), 1048-1052 (2000-12-01)
Fructose malabsorption is characterized by the inability to absorb fructose efficiently. As a consequence fructose reaches the colon where it is broken down by bacteria to short fatty acids, CO2 and H2. Bloating, cramps, osmotic diarrhea and other symptoms of
B P Chumpitazi et al.
Alimentary pharmacology & therapeutics, 42(4), 418-427 (2015-06-25)
A low fermentable oligosaccharides, disaccharides, monosaccharides and polyols (FODMAP) diet can ameliorate symptoms in adult irritable bowel syndrome (IBS) within 48 h. To determine the efficacy of a low FODMAP diet in childhood IBS and whether gut microbial composition and/or metabolic
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
Marine Zwicke et al.
Annals of botany, 116(6), 1001-1015 (2015-04-09)
Extreme climatic events such as severe droughts are expected to increase with climate change and to limit grassland perennity. The present study aimed to characterize the adaptive responses by which temperate herbaceous grassland species resist, survive and recover from a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service