Skip to Content
Merck
All Photos(3)

Documents

133612

Sigma-Aldrich

4-Biphenylacetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2CN
CAS Number:
Molecular Weight:
193.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

88-92 °C (lit.)

SMILES string

N#CCc1ccc(cc1)-c2ccccc2

InChI

1S/C14H11N/c15-11-10-12-6-8-14(9-7-12)13-4-2-1-3-5-13/h1-9H,10H2

InChI key

HSZCNGTZJWZAMF-UHFFFAOYSA-N

General description

4-Biphenylacetonitrile on condensation reaction with ethyl phenyl propiolate yields biphenyl derivatives of acetylenic β-ketocyanide.

Application

4-Biphenylacetonitrile was used to study the adsorption orientation of the biphenyl derivatives on the Au colloidal surfaces with α-cyclodextrins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensation of acetylenic esters with biphenyl compounds having active methylene groups.
Al-Jallo HN.
Journal of Chemical and Engineering Data, 17(4), 513-514 (1972)
The Fluorescence Study of the Induced Adsorption Orientation of Biphenyl Derivatives on Au Colloidal Surface.
Lee JK and Yu S-C.
Bull. Korean Chem. Soc., 30(7), 1637-1637 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service