Skip to Content
Merck
All Photos(1)

Key Documents

133450

Sigma-Aldrich

1-Amino-2,6-dimethylpiperidine

90%, technical grade

Synonym(s):

2,6-Dimethylpiperidin-1-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

refractive index

n20/D 1.465 (lit.)

bp

65-80 °C/30 mmHg (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

CC1CCCC(C)N1N

InChI

1S/C7H16N2/c1-6-4-3-5-7(2)9(6)8/h6-7H,3-5,8H2,1-2H3

InChI key

UAHWWAIVYPJROV-UHFFFAOYSA-N

Application

1-Amino-2,6-dimethylpiperidine was in the synthesis of N-substituted derivatives of 3-aminorhodanine.
Reactant for synthesis of:
  • Hydrazones
  • Superelectrophilic intermediates during 1,5-cyclization reactions
  • Amides
  • Anticonvulsant agents
  • HIV entry inhibitors for the CXCR4 receptor

Reactant for N-arylation reactions

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and antimicrobial activity of the 3-aminorhodanines.
ROWN FC, et al.
The Journal of Organic Chemistry, 24(8), 1056-1060 (1959)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service