Skip to Content
Merck
All Photos(1)

Key Documents

131911

Sigma-Aldrich

Tetrahydrofurfurylamine

97%

Synonym(s):

2-(Aminomethyl)tetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein:
102729
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

153-154 °C/744 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

NCC1CCCO1

InChI

1S/C5H11NO/c6-4-5-2-1-3-7-5/h5H,1-4,6H2

InChI key

YNOGYQAEJGADFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bolinaga R, et al.
The Journal of Chemical Thermodynamics, 10(7), 667-673 (1978)
Steffen Tröger-Müller et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(49), 11810-11817 (2017-05-05)
We report the synthesis of task-specific imidazolium ionic compounds and ionic liquids with key functionalities of organic molecules from electro-, polymer-, and coordination chemistry. Such products are highly functional and potentially suitable for technology applications even though they are formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service