Skip to Content
Merck
All Photos(1)

Key Documents

131415

Sigma-Aldrich

Phenyl 3-hydroxy-2-naphthoate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 104.00

CHF 104.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
CHF 104.00

About This Item

Linear Formula:
HOC10H6CO2C6H5
CAS Number:
Molecular Weight:
264.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 104.00


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

257-261 °C/160 mmHg (lit.)

mp

129-132 °C (lit.)

SMILES string

Oc1cc2ccccc2cc1C(=O)Oc3ccccc3

InChI

1S/C17H12O3/c18-16-11-13-7-5-4-6-12(13)10-15(16)17(19)20-14-8-2-1-3-9-14/h1-11,18H

InChI key

SRMZHGJLSDITLO-UHFFFAOYSA-N

Application

Phenyl 3-hydroxy-2-naphthoate was used in the synthesis of a series of new azoic coupling components, derivatives of naphthol AS[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of aizen spilon black TRH and its derivatives.
Dyes and Pigments, 14(4), 287-305 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service