Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.4326 (lit.)
bp
152-154 °C (lit.)
density
0.811 g/mL at 25 °C (lit.)
SMILES string
CCNCCNCC
InChI
1S/C6H16N2/c1-3-7-5-6-8-4-2/h7-8H,3-6H2,1-2H3
InChI key
CJKRXEBLWJVYJD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
N,N′-Diethylethylenediamine has been used as an internal standard in selective liquid chromatographic method developed for the assay of ethambutol in serum samples. It has also been used for the solvation of lithium hexamethyldisilazide.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
91.4 °F - closed cup
Flash Point(C)
33 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 20(3), 489-493 (2004-04-08)
A selective liquid chromatographic method has been developed for the assay of ethambutol in serum samples. The assay involves intramolecular excimer-forming derivatization with 4-(1-pyrene)butanoyl chloride (PBC) and isocratic reversed-phase chromatography with fluorescence detection. After acetonitrile-deproteinization of the serum sample, the
Dalton transactions (Cambridge, England : 2003), 46(13), 4452-4460 (2017-03-18)
In this study, crystalline magnesium chloride-electron donor complexes were prepared by recrystallization of δ-MgCl
Journal of the American Chemical Society, 118(14), 3529-3530 (1996)
Chemistry (Weinheim an der Bergstrasse, Germany), 23(64), 16231-16241 (2017-07-25)
The potential use of organoruthenium complexes as anticancer drugs is well known. Herein, a family of activatable tethered ruthenium(II) arene complexes of general formula [Ru{η
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service