Skip to Content
Merck
All Photos(2)

Documents

119261

Sigma-Aldrich

(Bicyclo[2.2.1]hepta-2,5-diene)tetracarbonylmolybdenum(0)

95%

Synonym(s):

(Norbornadiene)tetracarbonylmolybdenum, Bicyclo[2.2.1]heptadienemolybdenum tetracarbonyl, Tetracarbonyl(2,5-norbornadiene)molybdenum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8MoO4
CAS Number:
Molecular Weight:
300.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
core: molybdenum

mp

79-81 °C (lit.)

SMILES string

[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1C2C=CC1C=C2

InChI

1S/C7H8.4CO.Mo/c1-2-7-4-3-6(1)5-7;4*1-2;/h1-4,6-7H,5H2;;;;;

InChI key

UZHYHBPCAGKHGZ-UHFFFAOYSA-N

Application

The product may be used to modify multiwalled carbon nanotubes (MWCNTs) paste electrode with tetracarbonylmolybdenum for the determination of Cu(II) by square wave anodic stripping voltammetry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Square wave anodic stripping voltammetry of copper (II) at a MWCNT paste electrode modified with a tetracarbonylmolybdenum (0) nanocomposite.
Saidin MI, et al.
Microchimica Acta, 183.4, 1441-1448 (2016)
Unusual coordination of tetrylenes to molybdenum carbonyl fragments.
Lips F, et al.
Chemical Communications (Cambridge, England), 50.42, 5561-5564 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service