Skip to Content
Merck
All Photos(1)

Key Documents

111414

Sigma-Aldrich

1-Bromo-4-ethylbenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4Br
CAS Number:
Molecular Weight:
185.06
Beilstein:
1856989
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.544 (lit.)

bp

204 °C (lit.)

density

1.343 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(Br)cc1

InChI

1S/C8H9Br/c1-2-7-3-5-8(9)6-4-7/h3-6H,2H2,1H3

InChI key

URFPRAHGGBYNPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-4-ethylbenzene undergoes oxidation to give 4′-bromoacetophenone (74.3% yield).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-triethanolamine complex as efficient and active catalyst for selective oxidation of alkylarenes to phenyl ketones by tert-butylhydroperoxide.
Zhu M, et al.
Tetrahedron Letters, 48(52), 9108-9111 (2007)
Colin J Murphy et al.
Physical chemistry chemical physics : PCCP, 17(47), 31931-31937 (2015-11-17)
Surface-bound molecular rotation can occur with the rotational axis either perpendicular (azimuthal) or parallel (altitudinal) to the surface. The majority of molecular rotor studies involve azimuthal rotors, whereas very few altitudinal rotors have been reported. In this work, altitudinal rotors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service