Skip to Content
Merck
All Photos(1)

Key Documents

110183

Sigma-Aldrich

Potassium acetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOK
CAS Number:
Molecular Weight:
98.14
Beilstein:
3595449
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

solubility

water: soluble(lit.)

density

1.57 g/cm3 at 25 °C (lit.)

SMILES string

[K+].CC([O-])=O

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium acetate-catalyzed acetylation of wood: extraordinarily rapid acetylation at 120 C.
Obataya E and Minato K.
Wood Science and Technology, 42(7), 567-577 (2008)
Eagleson M.
Concise Encyclopedia Chemistry, 886-886 (1994)
Hendrik Marks et al.
Genome biology, 16, 149-149 (2015-08-04)
During early embryonic development, one of the two X chromosomes in mammalian female cells is inactivated to compensate for a potential imbalance in transcript levels with male cells, which contain a single X chromosome. Here, we use mouse female embryonic
Yuval Erez et al.
The journal of physical chemistry. A, 116(9), 2039-2048 (2012-02-10)
Steady-state and time-resolved emission techniques were employed to study the effect of acetate, a mild base, on the luminescence of curcumin in methanol and ethanol. We found that the steady-state emission intensity as well as the average fluorescence decay time
Susan Q Lang et al.
Rapid communications in mass spectrometry : RCM, 26(1), 9-16 (2012-01-05)
The stable isotopes of organic matter can provide valuable information on carbon cycling dynamics, microbial metabolisms, and past climates. Since bulk measurements may mask dynamic changes to critical portions of the organic pool, researchers are increasingly isolating individual compounds for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service