Skip to Content
Merck
All Photos(1)

Documents

110140

Sigma-Aldrich

Valeric acid

99%

Synonym(s):

n-Valeric acid, Pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
Beilstein:
969454
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)

Assay

99%

autoignition temp.

707 °F

expl. lim.

7.6 %

refractive index

n20/D 1.408 (lit.)

bp

110-111 °C/10 mmHg (lit.)
185 °C (lit.)

mp

−20-−18 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

CCCCC(O)=O

InChI

1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

NQPDZGIKBAWPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Böttcher et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 30(11), 1590-1596 (2000-11-09)
The prevalence of allergic diseases has increased particularly over the past 30-40 years. A reduced microbial stimulation during infancy may result in a development of a disturbed balance between Th1- and Th2-like immunity. The gut flora is, quantitatively, the most
D M Saunders et al.
The Australian & New Zealand journal of obstetrics & gynaecology, 18(3), 198-201 (1978-08-01)
The effect of oestradiol valerate on levels of blood cholesterol, triglycerides, free oestradiol and coagulation factors, as well as on urinary oestrogens and free cortisol was measured in 10 oöphorectomised women. Despite the very high urinary oestrogen levles obtained, there
A Gysler et al.
Pharmaceutical research, 16(9), 1386-1391 (1999-09-25)
To investigate pharmacokinetic differences between the nonhalogenated double ester prednicarbate (PC) and the fluorinated monoester betamethasone 17-valerate (BM17V) their metabolism in human keratinocytes and fibroblasts as well as their permeation and biotransformation in reconstructed epidermis and excised human skin was
Valeric biofuels: a platform of cellulosic transportation fuels.
Jean-Paul Lange et al.
Angewandte Chemie (International ed. in English), 49(26), 4479-4483 (2010-05-07)
Martin Koller et al.
Macromolecular bioscience, 7(2), 218-226 (2007-02-14)
Three different microbial wild-type strains are compared with respect to their potential as industrial scale polyhydroxyalkanoate (PHA) producers from the feed stock whey lactose. The halophilic archaeon Haloferax mediterranei as well as two eubacterial strains (Pseudomonas hydrogenovora and Hydrogenophaga pseudoflava)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service