Skip to Content
Merck
All Photos(1)

Key Documents

109843

Sigma-Aldrich

Phenol

reagent grade

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

reagent grade

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

autoignition temp.

1319 °F

expl. lim.

8.6 %

bp

182 °C (lit.)

mp

40-42 °C (lit.)

solubility

water: soluble 84 g/L at 20 °C

density

1.071 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyman JHP.
Synthetic and Natural Phenols., 67-67 (1996)
Eagleson M.
Concise Encyclopedia Chemistry, 812-812 (1994)
Tyman JHP.
Synthetic and Natural Phenols., 270-270 (1996)
Tyman JHP.
Synthetic and Natural Phenols., 1-1 (1996)
Phenol-hypochlorite reaction for determination of ammonia.
Weatherburn M W.
Analytical Chemistry, 39(8), 971-974 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service