Skip to Content
Merck
All Photos(2)

Key Documents

109150

Sigma-Aldrich

1,5-Dihydroxy-1,2,3,4-tetrahydronaphthalene

technical grade

Synonym(s):

1,2,3,4-Tetrahydro-1,5-naphthalenediol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 66.40

CHF 66.40


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
CHF 66.40

About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 66.40


Estimated to ship on22 May 2025


Request a Bulk Order

grade

technical grade

form

solid

mp

132-134 °C (lit.)

functional group

hydroxyl

SMILES string

OC1CCCc2c(O)cccc12

InChI

1S/C10H12O2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1,3,5,10-12H,2,4,6H2

InChI key

MYIDTCFDQGAVFL-UHFFFAOYSA-N

General description

1,5-Dihydroxy-1,2,3,4-tetrahydronaphthalene is a complexing agent and forms complexes with cephradine[1]. It reacts with 4- or 3-nitrobenzoyl chloride to yield 4-DABTN and 3-DABTN [2].

Application

1,5-Dihydroxy-1,2,3,4-tetrahydronaphthalene was used in the synthesis of 1,5-bis(4-aminobenzoate)-1,2,3,4-tetrahydronapthalene(4-DABTN) and 1,5-bis(3-aminobenzoate)-1,2,3,4-tetrahydronapthalene (5-DABTN)[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induced fit phenomena in clathrate structures of cephalosporins.
Kemperman GJ, et al.
Journal of the Chemical Society, 2(7), 1425-1429 (2000)
Synthesis of Novel Self-Developing Photosensitive Polyimide.
Ahn BH, et al.
Korea Polymer Journal, 8(1), 19-25 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service