Skip to Content
Merck
All Photos(1)

Key Documents

108499

Sigma-Aldrich

2-Bromododecane

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH(Br)CH3
CAS Number:
Molecular Weight:
249.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor pressure

0.29 psi ( 20 °C)

Assay

85%

refractive index

n20/D 1.456 (lit.)

bp

130 °C/6 mmHg (lit.)

mp

−96 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCCCCCCCCCC(C)Br

InChI

1S/C12H25Br/c1-3-4-5-6-7-8-9-10-11-12(2)13/h12H,3-11H2,1-2H3

InChI key

GIUUCQVKMWBSRT-UHFFFAOYSA-N

Application

2-Bromododecane was used as growth substrate to investigate the fatty acid composition of cellular lipids of Rhodococcus rhodochrous NCIMB 13064 grown on various long-chain haloalkanes[1]. It was used to compare the in vitro methods:yeast resazurin and 3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyltetrazolium bromide assay for determining the acute toxicity of halogenated alkanes[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of Yeast Resazurin Versus MTT Assay in vitro Methods For Determining Acute Toxicity of Halogenated Alkanes.
Zuas O.
Jurnal Ilmu Dasar, 9(2), 135-141 (2008)
Long-chain haloalkanes are incorporated into fatty acids by Rhodococcus rhodochrous NCIMB 13064.
Hamilton JTG, et al.
Microbiology, 141(10), 2611-2617 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service