Skip to Content
Merck
All Photos(1)

Key Documents

107948

Sigma-Aldrich

2-Methyl-3-hexanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COCH(CH3)2
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.406 (lit.)

bp

131-132 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

SMILES string

CCCC(=O)C(C)C

InChI

1S/C7H14O/c1-4-5-7(8)6(2)3/h6H,4-5H2,1-3H3

InChI key

HIGGFWFRAWSMBR-UHFFFAOYSA-N

Application

2-Methyl-3-hexanone was used in a study to investigate the natural abundance distribution of 1H and 2H in in sedimentary organic matter.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Equilibrium 2H/1H fractionations in organic molecules: I. Experimental calibration of ab initio calculations.
Wang Y, et al.
Geochimica et Cosmochimica Acta, 73(23), 7060-7075 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service