Z631167
Hettich® centrifuge buckets
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotors 1624; 1726
About This Item
Leistungsmerkmale
Holds 1 x 50 mLtube
Verpackung
pack of 6 ea
Hersteller/Markenname
Hettich 1746
Verfügbarkeit
available only in EU
not available in USA and Canada
Methode(n)
centrifugation: suitable
Kompatibilität
for use with rotors 1624 (Z601527) and 1726 (Z631108
Verwandte Kategorien
Allgemeine Beschreibung
- compatible with rotors 1624 (Z601527) and 1726 (Z631108)