Merck
TT38440SR-EP
Eppendorf twin.tec® PCR Plates 384
skirted, PCR clean, volume 40 μL, colorless wells, red plate, pack of 25
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41106300
eCl@ss:
32160702
Material:
colorless wells, polypropylene wells, red plate
Size:
384 wells
Sterility:
non-sterile
Feature:
skirt, PCR clean, skirted
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
colorless wells, polypropylene wells, red plate
sterility
non-sterile
feature
skirt, PCR clean, skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951020745 (NA)
technique(s)
PCR: suitable
size
384 wells
volume
40 μL
working volume
40 μL
General description
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Features and Benefits
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
  • 40 µL max. well volume
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
twin.tec is a registered trademark of Eppendorf SE
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)